Learn More
Invitrogen™ Human Apolipoprotein H Control Fragment Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ RP100507
Artikelnummer. 30196433
  • 248.00 CHF / 100 Mikroliter
Verfügbar ab: 06-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
P02749
Blocking Assay, Control
350
100 μL
RUO
APOH
Human
ECREVKCPFPSRPDNGFVNYPAKPTLYYKDKATFGCHDGYSLDGPEEIECTKLGNWSAMPSCKASCKVPVKKATVVYQGERVKIQEKFKNGMLHGDKVSFFCKNKEKKCSYTEDAQCIDGTIEVPKCFKEHSSLAFSKTDASDVK
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Apolipoprotein H
-20° C, Avoid Freeze/Thaw Cycles
2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG
Unconjugated
Recombinant
E. coli