Learn More
HMMR-Specific Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-15820-100UL
Additional Details : Gewicht : 0.00020kg
Beschreibung
The HMMR-Specific antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human HMMR-Specific. This antibody recognizes human, mouse, rat antigen. The HMMR-Specific antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Spezifikation
HMMR-Specific | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Flow Cytometry (Intracellular Staining) | |
CoraLite 488 | |
100 μL | |
3161 | |
IgG |