missing translation for 'onlineSavingsMsg'
Learn More

Histone H1 Rabbit, Clone: HH1/1784R, enQuireBio™

Rabbit Monoclonal Antibody

234.00 CHF - 523.00 CHF

Spezifikation

Antigen Histone H1
Klon HH1/1784R
Konzentration 200μg/mL
Anwendungen Immunohistochemistry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Quantity & Availability  
15960158
missing translation for 'viewDocuments'
EnquireBio
3005-RBM4-P0
20 μg Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A
15970158
missing translation for 'viewDocuments'
EnquireBio
3005-RBM4-P1
100 μg
523.00 CHF
100 Mikrogramm
Verfügbar ab: 25-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
Description

Description

Specifications

Specifications

Histone H1
200μg/mL
Monoclonal
Rabbit
10mM PBS with or without 0.05% BSA and 0.05% azide.
3005
Recombinant full-length human Histone H1 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
∽30kDa
HH1/1784R
Immunohistochemistry
Unconjugated
RUO
NP_005309
H1
IgG κ
Protein A/G Purified
Nuclear
HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.