Learn More
Heidolph™ Instruments Hei-VAP Powder Drying Package, CH-plug
The Hei-VAP Powder Drying Package features a motor lift, powder flask, and tubing set.
Marke: Heidolph™ Instruments 574-32300-04
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Hei-VAP Expert Control with motor lift and vertical glassware set G3
- Vacuum pump Rotavac Valve Control
- Vacuum valve
- Tubing set
- Powder flask 2 L
- Vapor tube with powder filter
Spezifikation
Motor Lift Rotary Evaporator | |
CH-plug |