Learn More
Heidolph™ Instruments Hei-VAP Automatic Evaporation Package, CH-plug
The Hei-VAP Automatic Evaporation Package features a motor lift, condenser, Woulff bottle, and tubing set.
Marke: Heidolph™ Instruments 574-37300-04
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Hei-VAP Expert Control with motor lift and vertical glassware set G3
- Vacuum pump Rotavac Vario Tec
- Condenser for Rotavac Vario
- Tubing set
- Woulff bottle
- AUTOaccurate-sensor
Spezifikation
Motor Lift Rotary Evaporator | |
CH-plug |