Learn More
Heidolph™ Hei-FLOW Expert 120
Der Pumpenantrieb des Modells 01 der Hei-FLOW Advantage Serie kann mit ein- oder mehrkanaligen Pumpenköpfen betrieben werden.
Marke:  Heidolph™ 523-51010-03
Artikelnummer. 17303450
  • 2975.00 CHF / 1 Stück
Verfügbar ab: 12-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • The flow rates with single-channel pump heads are in the range of 0.38 to 813 mL/min; with multichannel pump heads from 0.005 to 329 mL/min.
  • By integrating an analog interface, it is possible to control all functions externally. In addition, an optional foot switch can be adapted to enable dosing by foot pressure. Hands thus remain free to perform other work, for example, to hold the tubing and the vessel.
  • A speed of 5 to 120 rpm with a control accuracy of ± 3.5% guarantees optimum process results.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CH Plug
Analog
Variable-Speed Digital
230 V
Digitale variable Drehzahl
IP55
IP 55
Analog
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers