missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA588922
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: MPRYTVHVRG EWLAVPCQDA QLTVGWLGRE AVRRYIKNKP DNGGFTSVDD AHFLVRRCKG LGLLDNEDRL EVALENNEFV EVVIEGDAMS PDFIPSQPEG VYLYSKYREP EKYIELDGDR LTTEDLVNLG KGRYKIKLTP TAEKRVQKSR EVIDSIIKEK TVVYGITTGF GKFARTVIPI NKLQELQVNL VRSHSSGVGK; Positive Samples: Mouse liver, Rat liver
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.Spezifikation
HAL | |
Polyclonal | |
Unconjugated | |
HAL | |
hal, HIS, histidase, histidine ammonia lyase, histidine ammonia-lyase, histidinemia, Hsd, HSTD, Huth, zgc:136980 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15109, 29301, 3034 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.83 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
P21213, P35492, P42357 | |
HAL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HAL (NP_0020991) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |