missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
Antigen | GP repeat |
---|---|
Konzentration | 0.29 mg/mL |
Anwendungen | Immunohistochemistry, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16882965
|
Proteintech
24494-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
16872965
|
Proteintech
24494-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
This antibody is used to detect the GP repeated sequence. The recombit protein 6xHis tag-SMARCA4 or GST tag-SMARCA4 which contains the sequence 'GPGPGPGPG' were used to detect this antibody.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Spezifikation
GP repeat | |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C9RAN, GGGCC, GP repeat, GPGPGPGP, poly GP, poly PG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Peptide | |
Primary | |
-20°C |