Learn More
GFP tag Rabbit anti-Jellyfish (A. victoria), CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL594-50430-100UL
Additional Details : Gewicht : 0.00020kg
Beschreibung
The GFP tag antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of GFP tag. This antibody recognizes aequorea victoria, recombinant protein antigen. The GFP tag antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
GFP tag | |
Polyclonal | |
Rabbit | |
Primary | |
IgG |
Immunofluorescence | |
CoraLite 594 | |
100 μL | |
Jellyfish (A. victoria) |