Learn More
Biochrom Galapagos EZ Read PC Software
Galapagos EZ Read PC Software is intuitive, powerful and flexible software for all EZ Read Microplate Readers.
Marke: Biochrom 80-4002-41
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
You don't need to be an expert to use EZ Read PC software: our intuitive approach makes it easy to add analysis to your measurement.
Designed for Elisa Assays:
- Comprehensive formulae for built-in analysis of ELISA assays including curve-fitting options for 4 and 5 parameter fits-essential for many ELISA assays but difficult to do in Excel.
- Easy to configure your plate layout depending on the number of samples: add multiple assays to a plate for analysing just a few samples or add multiple plates for analysing.
Designed for Research Labs:
- Extensive offering of data analysis options for a range of absorbance assays.
- Downloadable assay library for quick method setup, just click and drag your samples and go.
Uncomplicated Setup for Measurements and Analysis:
- EZ Read PC software provides a clean and uncomplicated workspace to set up your analysis.
- Intuitive step by step approach for creating an integrated measurement and analysis method.
- Plate layout uses a simple 'click and drag' method for setting standard, control, sample and reference wells.
- Plate definition editor has preinstalled plates with the capability to modify for specific plates for best measurement performance.
Spezifikation
Microplate Reader | |
Software for Microplate Reader |