Learn More
Abnova™ FTH1 (Human) Recombinant Protein
Human FTH1 full-length ORF ( AAH11359, 1 a.a. - 183 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002495-P01.10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Sequence: MTTASTSQVRQNYHQDSEAAINRQINQELYASYVYLSMSYYFDRDDVALKNFAKYFLHQSHEEREHAEKLMKLQNQRGGRIFLQDIKKPDCDDWESGLNAMECALHLEKNVNQSLLELHKLATDKNDPHLCDFIETHYLNEQVKAIKELGDHVTNLRKMGAPESGLAEYLFDKHTLGDSDNES
Spezifikation
AAH11359 | |
ferritin, heavy polypeptide 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FTH1 | |
GST |
2495 | |
Wheat germ expression system | |
10 μg | |
FHC, FTH, FTHL6, MGC104426, PIG15, PLIF | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |