Learn More
Tocris Bioscience™ Fluorescein-NAD+
Fluorescent NAD+; substrate for ADP-ribosylation for use in PARP assays
Marke: Tocris Bioscience™ 6574/81U
Beschreibung
Fluorescein-NAD+ is a fluoroscein NAD+ is a substrate for ADP-ribosylation, providing a convenient non-isotopic alternative to radiolabelled NAD for use in PARP assays. Allows direct measurement of NAD-dependent enzymes, such as PARP, by fluorescence microscopy.This product is a replacement for R&D Systems product 4673-500-01
81 μg is supplied as 250 μl of a 0.25 mM solution in water.
Spezifikation
β-Nicotinamide-N6-[2-[[6-[fluorescein]-amino]-2-oxohexyl-hexyl]amino]-2-oxoethyl]adenine dinucleotide | |
81 μg | |
Enzyme Probes and Enzyme Substrates | |
C56H64N10O22P2 |