Learn More
Tocris Bioscience™ Fluorescein-NAD+
Fluorescent NAD+; substrate for ADP-ribosylation for use in PARP assays
Marke:  Tocris Bioscience™ 6574/81U
Artikelnummer. 18768436
  • 560.00 CHF / 81 Mikrogramm
Verfügbar ab: 22-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
Fluorescein-NAD+ is a fluoroscein NAD+ is a substrate for ADP-ribosylation, providing a convenient non-isotopic alternative to radiolabelled NAD for use in PARP assays. Allows direct measurement of NAD-dependent enzymes, such as PARP, by fluorescence microscopy.
This product is a replacement for R&D Systems product 4673-500-01
81 μg is supplied as 250 μl of a 0.25 mM solution in water.
Spezifikation
Spezifikation
β-Nicotinamide-N6-[2-[[6-[fluorescein]-amino]-2-oxohexyl-hexyl]amino]-2-oxoethyl]adenine dinucleotide
81 μg
Enzyme Probes and Enzyme Substrates
C56H64N10O22P2