Learn More
Proteintech FlexAble CoraLite Plus 405 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 405 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 405 in any buffer condition to Human IgG primary antibodies from any supplier.
Marke: Proteintech KFA108-200
Additional Details : Gewicht : 0.02000kg
Beschreibung
FlexAble CoraLite™ Plus 405 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 405 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
100 Tests | |
Liquid |