Learn More
Proteintech FlexAble Biotin Antibody Labeling Kit for Human IgG
FlexAble Biotin Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate Biotin in any buffer condition to Human IgG primary antibodies from any supplier.
102.24 CHF - 1009.87 CHF
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18079365
|
Proteintech
KFA111-10 |
10 Tests |
102.24 CHF
10 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18035505
|
Proteintech
KFA111-200 |
100 Tests |
1009.87 CHF
200 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18018075
|
Proteintech
KFA111-50 |
50 Tests |
475.97 CHF
50 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
FlexAble Biotin Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate Biotin in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.