Learn More
F(ab')2-Rabbit anti-Rat IgG (H+L) Secondary Antibody, AP, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA129928
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA1-29928 detects Rat IgG (H+L) from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Rat IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
purified | |
RUO | |
Rat | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
0.05M tris HCl with 0.15M NaCl, 0.1mM ZnCl2, 10mg/mL BSA, 1mM MgCl2, 50% glycerol and 0.05% sodium azide; pH 8 | |
Rat IgG whole molecule | |
250 μg | |
Secondary | |
4° C, do not freeze | |