Learn More
Invitrogen™ Dynabeads™ Sheep-Anti Mouse IgG
Dynabeads™ Sheep-Anti Mouse IgG
Marke:  Invitrogen™ 11031
Artikelnummer. 15474683
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.
  • Depletion: cells can be depleted from a sample
  • Positive isolation: cells can be positively isolated from a sample for molecular applications
  • Negative isolation: cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.
    Note: For a more universal binding of your mouse IgG of any subclass, please use Dynabeads™ Pan Mouse Ig (Cat.no. 110-41D or 110-42D).
    Starting samples:
    Whole blood, PBMC, buffy coat., tissue digests.
    TRUSTED_SUSTAINABILITY
  • Spezifikation
    Spezifikation
    2 °C bis 8 °C
    Dynabeads™ Sheep-Anti Mouse IgG, 5mL, ∽2 x 10^9 Cells Total Capacity, 1 x 10^7 PBMCs per Isolation, Polyclonal, Sheep Host, Whole Antibody Fragment, >95% Output Viability, 7.4 pH, PBMC, Blood, Tissue Digests Sample, Used for Cell Isolation a
    Dynabeads™
    Forschungsqualität
    Blood, PBMC, tissue digests
    5mL Dynabeads™ sheep anti-mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3)
    RNAi, Epigenetics and non-coding RNA Research, Chromatin Biology, Cell Analysis, Cell Isolation, Cell Expansion, ChIP
    1 Each
    5 mL
    Stable