Learn More
Invitrogen™ Dynabeads™ Sheep-Anti Mouse IgG
Dynabeads™ Sheep-Anti Mouse IgG
Marke: Invitrogen™ 11031
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.Note: For a more universal binding of your mouse IgG of any subclass, please use Dynabeads™ Pan Mouse Ig (Cat.no. 110-41D or 110-42D).
Starting samples:
Whole blood, PBMC, buffy coat., tissue digests.
Spezifikation
2 °C bis 8 °C | |
Dynabeads™ Sheep-Anti Mouse IgG, 5mL, ∽2 x 10^9 Cells Total Capacity, 1 x 10^7 PBMCs per Isolation, Polyclonal, Sheep Host, Whole Antibody Fragment, >95% Output Viability, 7.4 pH, PBMC, Blood, Tissue Digests Sample, Used for Cell Isolation a | |
Dynabeads™ | |
Forschungsqualität | |
Blood, PBMC, tissue digests |
5mL Dynabeads™ sheep anti-mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3) | |
RNAi, Epigenetics and non-coding RNA Research, Chromatin Biology, Cell Analysis, Cell Isolation, Cell Expansion, ChIP | |
1 Each | |
5 mL | |
Stable |
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.