Learn More
Elma™ Drain Sieve for Parts Holder
Stainless steel sieve for use in the drain of an Elmasonic ultrasonic device of size 30 to 300.
Marke: Elma™ 1045648
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Dirt particles ≥0.15 mm are caught in the wire mesh of the drain strainer and thus cannot clog the water drain
- Clean items with a wide variety of geometries, from small watch components and items of jewelry to conical flasks, test tubes and even dental products in your ultrasonic bath
- Optimum positioning of the items inside the bath is a prerequisite for achieving the best possible cleaning results
- Allows items to be individually positioned
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
- Holders are available for various applications
- Special solutions are available (e.g. for conical flasks)
Spezifikation
Drain Sieve | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |