Learn More
Aalborg™ MASSFLOW METER DIGITAL 0-5L/MIN
Aalborg™ Digital Thermal Mass Flow Controller allows one to program, record, and analyze flow rates of various gases with a computer.
Marke:  Aalborg™ DFCS-011428
Artikelnummer. 13427246
  • 2815.00 CHF / 1 Stück
Verfügbar ab: 13-03-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Digital and analog modes operate
  • Programmable flow configurations
  • Available flow ranges starting from 0 to 10 sccm up to 0 to 100 slpm
  • Maximum pressure of 1000 psig (70 bars)
  • Multi-drop capability of up to 256 units
  • Stores calibration data for up to 10 gases
  • Totalizer indicates total gas quantity
  • Alarm limits for high and low gas flow
  • Conversion factors for up to 256 gases
  • Auto tune function for optimum control response
  • Self diagnostic test
Spezifikation
Spezifikation
Digital Regulator
5 LPM
70 bar
0 bis 5 LPM