Learn More
Novus Biologicals™ Cytochrome b reductase 1 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-09660
Artikelnummer. 18287032
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 23-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Cytochrome b reductase 1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC29455 and based on accession number NM_24843. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens cytochrome b reductase 1 (CYBRD1), transcript variant 1, mRNA.
Native
Sonderangebote
Nur für Forschungszwecke