Learn More
Novus Biologicals™ CSTP1 Overexpression Lysate
Marke: Novus Biologicals™ NBP2-06819
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CSTP1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_681. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Western Blot | |
RIPA buffer | |
Homo sapiens CSTP1, mRNA. | |
Signaltransduktion |
Nur für Forschungszwecke