Learn More
CREST Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech CL594-12439-100UL
Additional Details : Gewicht : 0.00010kg
Artikelnummer. 17264776
  • 350.75 CHF / 100 Mikroliter
Verfügbar ab: 19-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CREST, also named as SS18-like 1(SS18L1) is a transcriptional activator that is required for calcium-dependent dendritic growth and branching in cortical neurons. It's also a nuclear protein interacts with CREB-binding protein and expressed in the developing brain. It helps regulate neuronal morphogenesis in calcuim-dependent manner. The N-terminal domain of SS18L1 is required for suppressing transactivation in the basal state, while the C-terminal domain is required for calcium-induced transactivation. It's part of the CREST-BRG1 complex, a multiprotein complex that regulates promoter activation by orchestrating a calcium-dependent release of a repressor complex and a recruitment of an activator complex. This complex also binds to the NR2B promoter, and activity-dependent induction of NR2B expression involves a release of HDAC1 and recruitment of CREBBP. The calculated molecular weight of CREST is about 43 kDa, but the modified of CREST protein is 55 kDa (PMID: 25888396). CREST exists some isoforms with calulated MV 43, 40, 33 and 29 kDa.
Spezifikation
Spezifikation
CREST
Polyclonal
CoraLite 594
100 μL
26039
Liquid
Immunofluorescence
1000 μg/mL