Learn More
Kupfer-Etzanchant, Thermo Scientific Chemicals
349.00 CHF
Spezifikation
Chemischer Name oder Material | Copper etchant |
---|---|
Dichte | 1.19 |
MDL-Nummer | MFCD00161912 |
Form | Flüssig |
Beschreibung
Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Copper etchant | |
1.19 | |
MFCD00161912 | |
Flüssig |
Sicherheit und Handhabung
P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P390-P501c
H290-H302-H315-H317-H318
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'tsca' : Yes
Recommended Storage : Umgebungstemperaturen
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.