Learn More
COOMBS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA17678
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA1-7678 detects COOMBS protein in human samples. PA1-7678 has been successfully used in agglutination applications. The PA1-7678 immunogen is a COOMBS protein.
COOMBS Polyclonal antibody specifically detects COOMBS in Human samples. It is validated for Agglutination.Spezifikation
COOMBS | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |
Agglutination | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
COOMBS. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |