Learn More
Novus Biologicals™ Connexin 25/GJB7 Overexpression Lysate
Marke: Novus Biologicals™ NBL1-11099
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Connexin 25/GJB7 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC29282 and based on accession number NM_198568. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens gap junction protein, beta 7, 25kDa (GJB7), mRNA. | |
Native |
Nur für Forschungszwecke