missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP164 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22227-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CEP164 was initially identified as a centrosomal protein, but other studies have indicated that it also plays a role in the formation of primary cilia, the microtubule-based sensory antennae projecting from the surface of many eukaryotic cells as well as in DNA damage response acting as a mediator protein. CEP164 interacts with both ATR and ATM, proteins that trigger a number of cellular responses including the initiation of DNA damaged-induced cell cycle checkpoints. It is phosphorylated upon replication stress, ultraviolet (UV) radiation, and ionizing radiation silencing of CEP164 significantly reduces the DNA damage-induced phosphorylation of several proteins in the DNA damage-activated signaling cascade and compromises cell survival after UV damage. At least two isoforms of CEP164 are known to exist.Spezifikation
CEP164 | |
Polyclonal | |
Unconjugated | |
CEP164 | |
centrosomal protein 164kDa, Centrosomal protein of 164 kDa, CEP164, KIAA1052 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22897, 489389 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UPV0 | |
CEP164 | |
CEP164 Fusion Protein Ag17570 | |
150 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG |