missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK2/cyclin O Recombinant Human Protein

CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. Recombinant full-length human CDK2/Cyclin O was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

574.00 CHF - 8710.00 CHF

Spezifikation

Zur Verwendung mit (Anwendung) Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
Zusammensetzung 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
Molekulargewicht 60.2/64.4kDa
Lagerungsbedingungen Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Gen-Alias CDK2: p33 (CDK2) CyclinO: CCNO, CCNU, UDG2
Mehr Spezifikationen ansehen

Produkte 3
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15478933
Unterlagen ansehen
Invitrogen™
PV6286
10 μg
574.00 CHF
10 Mikrogramm
Verfügbar ab: 04-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15488933
Unterlagen ansehen
Invitrogen™
PV6287
100 μg
1942.00 CHF
100 Mikrogramm
Verfügbar ab: 04-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15498933
Unterlagen ansehen
Invitrogen™
PV6288
1 mg
8710.00 CHF
1 Milligramm
Verfügbar ab: 04-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Spezifikation

Spezifikation

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
60.2/64.4kDa
CDK2: p33 (CDK2) CyclinO: CCNO, CCNU, UDG2
Recombinant
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
Videos
SDS
Documents

Documents

Certificates