Learn More
CD62P Mouse anti-Human, PE, Clone: AK-4, BD
Mouse Monoclonal Antibody
114.00 CHF - 255.00 CHF
Spezifikation
Antigen | CD62P |
---|---|
Klon | AK-4 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15861469
|
BD Biosciences
561921 |
25 Tests |
114.00 CHF
1 Stück |
Verfügbar ab: 18-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10242483
|
BD Biosciences
555524 |
100 Tests |
255.00 CHF
1 Stück |
Verfügbar ab: 18-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.
Spezifikation
CD62P | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
AK-4 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
P-Selectin; GMP-140; PADGEM |
For Research Use Only.