missing translation for 'onlineSavingsMsg'
Learn More

CD59 (Protectin) Recombinant Rabbit Monoclonal Antibody (JM10-71), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen MA532588

Artikelnummer. 16385994

  • 604.00 CHF / 100 Mikroliter
Verfügbar ab: 17-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

CD59 (Protectin)
Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4
O55186, P13987, P58019
CD59, Cd59a, Cd59b
Synthetic peptide within Human CD59 aa 85-121
100 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, Western Blot
JM10-71
Unconjugated
CD59
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, AA987121, Cb59b molecule, CD59, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344), CD59 glycoprotein, CD59 molecule, CD59 molecule (CD59 blood group), CD59 molecule, complement regulatory protein, Cd59a, CD59a antigen, CD59A glycoprotein, Cd59b, CD59b antigen, CD59B glycoprotein, CD59b moleucle, complement regulatory protein, complement inhibitor CD59, complement regulatory protein, EJ16, EJ30, EL32, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MACIP, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20, protectin, surface anitgen recognized by monoclonal antibody 16.3A5, T cell-activating protein
Rabbit
Protein A
RUO
12509, 333883, 966
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt