missing translation for 'onlineSavingsMsg'
Learn More

CD59 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

162.00 CHF - 344.00 CHF

Spezifikation

Antigen CD59
Konzentration 0.5 mg/mL
Anwendungen Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17948942
Unterlagen ansehen
Invitrogen
PA5145340
20 μL
162.00 CHF
20 Mikroliter
Verfügbar ab: 23-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
17969602
Unterlagen ansehen
Invitrogen
PA5145339
200 μL
344.00 CHF
200 Mikroliter
Verfügbar ab: 23-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.
Spezifikation

Spezifikation

CD59
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse
O55186, P13987, P58019
12509, 333883, 966
Recombinant Mouse CD59 protein, Val22-Ser96 (Accession #O55186)
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, AA987121, Cb59b molecule, CD59, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344), CD59 glycoprotein, CD59 molecule, CD59 molecule (CD59 blood group), CD59 molecule, complement regulatory protein, Cd59a, CD59a antigen, CD59A glycoprotein, Cd59b, CD59b antigen, CD59B glycoprotein, CD59b moleucle, complement regulatory protein, complement inhibitor CD59, complement regulatory protein, EJ16, EJ30, EL32, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MACIP, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20, protectin, surface anitgen recognized by monoclonal antibody 16.3A5, T cell-activating protein
CD59, Cd59a, Cd59b
IgG
Protein A
CD59
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt