Learn More
Novus Biologicals™ CD59 Overexpression Lysate
Marca: Novus Biologicals™ NBP2-09435
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD59 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifica
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_112699. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Western Blot | |
RIPA buffer | |
Homo sapiens CD59, mRNA. | |
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein |
Nur für Forschungszwecke