Learn More
CD14 Mouse anti-Human, FITC, Clone: UCHM-1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech FITC-65056-100TESTS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with thymocytes and peripheral blood lymphocytes from a remission aml patient of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Spezifikation
CD14 | |
Monoclonal | |
10 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
UCHM-1 | |
FITC | |
100 Tests | |
929 | |
IgG2a |