Learn More
CD14 Mouse anti-Human, FITC, Clone: UCHM-1, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech FITC-65056-100TESTS
Artikelnummer. 16821176
  • 59.30 CHF / 100 Tests
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with thymocytes and peripheral blood lymphocytes from a remission aml patient of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Spezifikation
Spezifikation
CD14
Monoclonal
10 μL/Test
Mouse
Primary
Human
Flow Cytometry
UCHM-1
FITC
100 Tests
929
IgG2a