missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b/c Mouse anti-Rat, FITC, Clone: OX-42, BD
Mouse Monoclonal Antibody
170.00 CHF - 820.00 CHF
Spezifikation
Antigen | CD11b/c |
---|---|
Klon | OX-42 |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15861239
|
BD Biosciences
561691 |
50 μg |
170.00 CHF
1 Stück |
Verfügbar ab: 20-09-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10089333
|
BD Biosciences
554861 |
0.5 mg |
820.00 CHF
1 Stück |
Verfügbar ab: 20-09-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The OX-42 monoclonal antibody specifically binds to the CR3 complement (C3bi) receptor found on most monocytes, granulocytes, macrophages, dendritic cells, and microglia. It appears to recognize a common epitope shared by CD11b and CD11c (integrin αM and αX chains). OX-42 antibody inhibits C3bi binding activity.
Spezifikation
CD11b/c | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Rat | |
IgG2a κ | |
Affinity Purified |
OX-42 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.