Learn More
CASK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1544
Beschreibung
PA1-544 detects calmodulin sensitive kinase (CASK) from mouse and rat tissues and cells. PA1-544 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects a 112 kDa protein representing CASK from AtT20 cell lysate. Immunohistochemical staining of CASK in mouse brain with PA1-544 results in staining of the choroid plexus. PA1-544 immunizing peptide corresponds to amino acid residues 578-592 from human CASK. This sequence is completely conserved between rat, mouse, and human CASK. PA1-544 can be used with blocking peptide PEP-110.
CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.Spezifikation
CASK | |
Polyclonal | |
Unconjugated | |
Cask | |
CAGH39; calcium/calmodulin dependent serine protein kinase; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; G-protein-coupled receptor GPR34; hCASK; LIN2; LIN-2; MGC7449; MICPCH; mLin-2; MRXSNA; OTTHUMP00000023157; OTTHUMP00000023161; OTTHUMP00000023162; Pals3; peripheral plasma membrane protein CASK; Protein lin-2 homolog; RP23-278L4.1; TNRC8; trinucleotide repeat containing 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12361, 29647, 8573 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O14936, O70589, Q62915 | |
Cask | |
Synthetic Peptide: C(578) E R D S P S T S R Q S P A N(592) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.