Learn More
CASK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA596141
Beschreibung
Immunogen sequence: MADDDVLFED VYELCEVIGK GPFSVVRRCI NRETGQQFAV KIVDVAKFTS SPGLSTEDLK REASICHMLK HPHIVELLET YSSDGMLYMV FEFMDGADLC FEIVKRADAG FVYSEAVASH YMRQILEALR YCHDNNIIHR DVKPHCVLLA SKENSAPVKL GGFGVAIQLG ESGLVAGGRV GTPHFMAPEV VKREPYGKPV DVWGCGVILF ILLSGCLPFY GTKERLFEGI IKGKYKMNPR QWSHISESAK DLVRRMLMLD PAERITVYEA LNHPWLKERD RYAYKIHLPE TVEQLRKFNA; Positive Samples: 293T, U-251MG, BxPC-3, 22Rv1, NCI-H460, A-549; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Peripheral membrane protein
CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.Spezifikation
CASK | |
Polyclonal | |
Unconjugated | |
Cask | |
CAGH39, calcium/calmodulin dependent serine protein kinase, calcium/calmodulin-dependent serin protein kinase, Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase, CAMGUK, CASK, CMG, DXPri1, DXRib1, FGS4, G-protein-coupled receptor GPR34, hCASK, LIN2, LIN-2, MGC7449, MICPCH, mLin-2, MRXSNA, OTTHUMP00000023157, OTTHUMP00000023161, OTTHUMP00000023162, Pals3, peripheral plasma membrane protein CASK, Protein lin-2 homolog, RP23-278L4.1, TNRC8, trinucleotide repeat containing 8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8573 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
O14936 | |
Cask | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CASK (NP_0011195261) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.