missing translation for 'onlineSavingsMsg'
Learn More

Kohlenstoffdisulfid, niedriges Benzol, 99.9 %, Thermo Scientific Chemicals

Artikelnummer. 15411677
Click to view available options
Menge:
100 mL
500 mL
4 x 500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates.

Carbon disulfide is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reactive agent for the production of rubber vulcanization accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 75-15-0
Summenformel CS2
Molekulargewicht (g/mol) 76.131
MDL-Nummer MFCD00011321
InChI-Schlüssel QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC-Name Methandithion
SMILES C(=S)=S

Spezifikation

Chemischer Name oder Material Carbon disulfide
Schmelzpunkt °C to 112°C
Dichte 1.2632 g/mL
Siedepunkt 46°C
Menge 4 x 500 mL
Flammpunkt -30°C (-22°F)
Physikalische Form Flüssig
Brechungsindex 1.627
Prozentgehaltsbereich 99.9%
Löslichkeitsinformationen Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formelmasse 76.14
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'