Learn More
R&D Systems™ Canine HGFR/c-MET ECD DuoSet ELISA
Sufficient for at least 15 96-well plates
Marke: R&D Systems™ DY4140
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Canine HGF R/c-MET ECD DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies canine HGFR/c-MET.Spezifikation
Biotin | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
156.00 to 10000 pg/mL | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |