Learn More
Abnova™ CALM3 (Human) Recombinant Protein
Human CALM3 full-length ORF ( AAH05137.1, 1 a.a. - 149 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00000808-P01.25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Sequence: MADQLTEEQIAEFKEAFSLFDKDGDGTITTKELGTVMRSLGQNPTEAELQDMINEVDADGNGTIDFPEFLTMMARKMKDTDSEEEIREAFRVFDKDGNGYISAAELRHVMTNLGEKLTDEEVDEMIREADIDGDGQVNYEEFVQMMTAK
Spezifikation
AAH05137.1 | |
calmodulin 3 (phosphorylase kinase, delta) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CALM3 | |
GST |
808 | |
Wheat germ expression system | |
25 μg | |
PHKD, PHKD3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |