missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C2C12 Whole Cell Lysate
C2C12 (muscle, myoblast) whole stem cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: DMEM and 10% FBS (fetal bovine serum). Lysates are guaranteed for 6 months from date of receipt.
Marke: Novus Biologicals™ NBP2-10268-0.1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The C2C12 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.Spezifikation
Western Blot | |
0.1 mg | |
C2C12 (muscle; myoblast) | |
2 mg/mL |
C2C12 | |
Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. | |
Mouse | |
Store at −80°C. Avoid freeze-thaw cycles. |