missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan Brinometer Hydrometer
Special purpose hydrometer to measure the precentage of saturated solution of brine in water at 15.6°C
Marke: Brannan 10/621/0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- 0 to 100% range
- 230 mm long and 1% adjusted at 15.6°C
- Made in UK
- Supplied in protective, resealable plastic tube
Spezifikation
1 | |
±1.0 | |
23 cm | |
Adjusted at 15.6°C | |
Stahlschrot |
Hydrometer | |
0.1 | |
Protective Plastic Tube | |
Soda Glass |