Learn More
Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
Marke:  Novus Biologicals™ NBP2-47118
Artikelnummer. 18163269
  • 542.10 CHF / 2 Stück
Verfügbar ab: 04-11-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
These lysates have not been subjected to denaturing or reducing conditions. This allows the tissue or cell lysate to be used in a variety of applications; to study protein-protein interaction, ligand binding, ELISA, immunoprecipitation, 1D and 2D gel electrophoresis, and Western blotting for the detection of specific protein targets. For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required. Buffer requirements for performing protein-protein interaction and ligand binding studies can vary significantly from RIPA buffer and may require modifications. In most cases, tissue lysates in RIPA buffer can be used, directly in standard ELISA and immunoprecipitation assays. These lysates are proteomic discovery tools. Researchers should validate and optimize for individual use.
The Breast Matched Tumor & Normal Tissue Lysate is derived from human.
Spezifikation
Spezifikation
Breast
2 Fläschchen
Human
Store at −80°C. Avoid freeze-thaw cycles.