Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Gas Washing Bottle without Filter Disc
Brand: Lenz Laborglasinstrumente™ 05500558
This item is not returnable.
View return policy
Specifications
500 mL | |
Gas Washing Bottle | |
NS 29/32 | |
1 Pack |
Borosilicate Glass, Duran Tubing | |
Head | |
DIN 12596 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Gas Washing Bottle without Filter Disc > Capacity: 500mL
Spot an opportunity for improvement?Share a Content Correction