Learn More
Brady™ BMP™61 Series Printer Ribbon - R6000 Resin, Black
BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.
Marke: Brady™ 148128
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- One-color ribbon for printing a single color onto a label
- R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material
- Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
- Automatic label formatting with smart cell technology and easy loading
Spezifikation
Black |