Learn More
Benchmark Scientific™ Block for Centrifuge Tubes
Click to view available options
Benchmark Scientific™ Conical Microtube Holder
Beschreibung
- Accommodates 15 x 5 mL conical microtubes
- Protective shield protects users from burns
- Fits MultiTherm H and HC Models
Spezifikation
Autoklavierbar | Not Autoclavable |
Kammermaterial | Aluminum |
Aufnahmevermögen | 15 x 5 mL Microtubes, Conical |
Material | Aluminum |
No. of Places | 15 |
Typ | Block |