Learn More
Brand™ BLAUBRAND™ ETERNA™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper, with Certificate
Borosilicate glass construction and calibrated to contain (TC, In.) Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. Delivered with individual USP conformity certificate.
Marke: Brand™ 956841
52.40 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Borosilicate glass (Boro 3.3) construction
- Polypropylene stopper
- Graduation markings in amber
- Calibrated to contain (TC, In)
- DIN EN ISO 1042
- Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
- Delivered with individual USP conformity certificate
Spezifikation
Calibrated To Contain | |
Polypropylen | |
25 mL | |
±0.04 ml | |
Yes | |
10/19 |
USP-Einzel- und USP-Chargenzertifikat, DIN EN ISO 1042 | |
Klar | |
Borosilicate Glass | |
2 Stück | |
A |