Learn More
Bel-Art™ SP Scienceware™ General Purpose Trays Without Faucets
Rugged, long-lasting and corrosion resistant plastic
153.00 CHF
Beschreibung
- All trays have heavy walls and wide top flanges
- Molded from low-density polyethylene
- Withstand temperatures up to 77°C (171°F)
Spezifikation
Polyethylen | |
Tablett |