Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Chloramphenicol29
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
47.50 CHF
Spezifikation
Antimikrobielle Mittel | Chloramphenicol29 |
---|---|
Verpackungsart | Scheibe |
Beschreibung
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Spezifikation
Chloramphenicol29 | |
Scheibe |
For in vitro diagnostic use