missing translation for 'onlineSavingsMsg'
Learn More

Bariumdodecaeisennonadecaoxid, Thermo Scientific Chemicals

Artikelnummer. 15432357
Click to view available options
Menge:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Barium dodecairon nonadecaoxide is used in subwoofer magnets to provide power. It is also used in new high tech magnetic tapes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 12047-11-9
Summenformel BaFe12O19
MDL-Nummer MFCD00075637
Synonym Barium ferrite

Spezifikation

Chemischer Name oder Material Barium dodecairon nonadecaoxide
Schmelzpunkt >1315°C
Farbe Schwarz
Physikalische Form Pulver
Menge 100 g
UN-Nummer UN1564
Löslichkeitsinformationen Insoluble in water.
Formelmasse 1111.48

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'