Learn More
AZ PFKFB3 26, Tocris Bioscience™
Artikelnummer. 30002174
Click to view available options
Menge:
10 mg
50 mg
Potent and selective PFKFB3 inhibitor
Potent and selective PFKFB3 inhibitor (IC50 = 0.023 μM). Selective for PFKFB3 over PFKFB2 and PFKFB1 isoforms (IC50 values are 0.382 μM and 2.06 μM, respectively). Reduces levels of fructose-1,6-bisphosphate in A549 cells (IC50 = 0.343 μM). Exhibits no significant inhibition of other kinases.
Chemische Identifikatoren
CAS 1704740-52-2
Summenformel C24H26N4O2
Molekulargewicht (g/mol) 402.498
InChI-Schlüssel OOGHGWKBJXQNEJ-QFIPXVFZSA-N
Synonym az pfkfb3 26, 2s-n-4-3-cyano-1-2-methylpropyl-1h-indol-5-yl oxy phenyl-2-pyrrolidinecarboxamide
PubChem CID 118735769
IUPAC-Name (2S)-N-[4-[3-cyano-1-(2-methylpropyl)indol-5-yl]oxyphenyl]pyrrolidine-2-carboxamide
SMILES CC(C)CN1C=C(C2=C1C=CC(=C2)OC3=CC=C(C=C3)NC(=O)C4CCCN4)C#N
Spezifikation
Chemischer Name oder Material (2S)-N-[4-[[3-Cyano-1-(2-methylpropyl)-1H-indol-5-yl]oxy]phenyl]-2-pyrrolidinecarboxamide
Strukturformel C24H26N4O2
Menge 10 mg
Löslichkeitsinformationen Soluble to 100mM in DMSO and to 100mM in ethanol
Formelmasse 402.49
Reinheit (%) >98%
Physikalische Form Solid