Learn More
AZ 628, Tocris Bioscience™
Artikelnummer. 30001584
Click to view available options
Menge:
10 mg
50 mg
Potent Raf kinase inhibitor
Potent, ATP-competitive inhibitor of Raf kinases (IC50 values are 29, 34 and 105 nM for c-Raf1, B-RafV600E and wild-type B-Raf, respectively). Displays selectivity for Raf kinases over a panel of 150 other kinases; inhibits activation of tyrosine protein kinases such as VEGFR2, Lyn, Flt1 and Fms. Also inhibits growth, and induces cell cycle arrest and apoptosis in colon and melanoma cell lines with the B-RafV600E mutation.
Chemische Identifikatoren
CAS 878739-06-1
Summenformel C27H25N5O2
Molekulargewicht (g/mol) 451.53
MDL-Nummer MFCD17392577
InChI-Schlüssel ZGBGPEDJXCYQPH-UHFFFAOYSA-N
Synonym 3-2-cyanopropan-2-yl-n-4-methyl-3-3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl amino phenyl benzamide, 3-1-cyano-1-methylethyl-n-3-3,4-dihydro-3-methyl-4-oxo-6-quinazolinyl amino-4-methylphenyl benzamide, 3-2-cyanopropan-2-yl-n-4-methyl-3-3-methyl-4-oxo-3,4-dihydroquinazolin-6-ylamino phenyl benzamide, 3-2-cyanopropan-2-yl-n-4-methyl-3-3-methyl-4-oxoquinazolin-6-yl amino phenyl benzamide, 3-1-cyano-1-methylethyl-n-4-methyl-3-3-methyl-4-oxoquinazolin-6-yl amino phenyl benzamide, b1e, benzamide, 3-1-cyano-1-methylethyl-n-3-3,4-dihydro-3-methyl-4-oxo-6-quinazolinyl amino-4-methylphenyl, 3-1-cyano-1-methylethyl-n-4-methyl-3-3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl amino phenyl benzamide
PubChem CID 11676786
IUPAC-Name 3-(1-cyano-1-methylethyl)-N-{4-methyl-3-[(3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino]phenyl}benzamide
SMILES CN1C=NC2=CC=C(NC3=C(C)C=CC(NC(=O)C4=CC(=CC=C4)C(C)(C)C#N)=C3)C=C2C1=O
Spezifikation
Chemischer Name oder Material AZ 628
Menge 10 mg
Formelmasse 451.52
Reinheit (%) >98%