missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoptosis-Enhancing Nuclease Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA5111279
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: SLTIPNAKDV LRKRHKRRSR QHQRFMARKA LLQEQGLLSM PPEPGSSPLP TPFGAATATE AASSGKQCLR AGSGSAP
Apoptosis-Enhancing Nuclease Polyclonal antibody specifically detects Apoptosis-Enhancing Nuclease in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Apoptosis-Enhancing Nuclease | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WTP8 | |
AEN | |
Recombinant Protein corresonding to Human Apoptosis-Enhancing Nuclease | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
AEN | |
2700083B06Rik; AEN; aen {ECO:0000250; apoptosis enhancing nuclease; apoptosis-enhancing nuclease; apoptosis-enhancing nuclease {ECO:0000250; interferon stimulated exonuclease gene 20kDa-like 1; interferon stimulated exonuclease gene 20-like 1; interferon-stimulated 20 kDa exonuclease-like 1; interferon-stimulated 20 kDa exonuclease-like 1 {ECO:0000312; ISG20L1; pp12744; RGD:1305051}; RGD1305051; SBBI58; UniProtKB:Q8WTP8} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64782 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |